4-Chlor-3-sulphamoylbenzoicsyre

Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PROOMM0477.04EA 5530 DKK
PROOMM0477.04
4-Chlor-3-sulphamoylbenzoicsyre
4-Chlor-3-sulphamoylbenzoicsyre
Formel: C₇H₆ClNO₄S
Molekylvægt: 235,65 g/mol
MDL Number: MFCD00012375
CAS-nummer: 1205-30-7
EINECS: 214-882-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR