Hotplates RC series
Leverandør: VELP Scientifica
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
453-0145EA 3950 DKK
453-0145
Hotplates RC series
Varmeapparater Varmeplader
Hotplates with coated aluminium alloy plates give even heat distribution. Choice of models with one or two plates.
  • Housing has good chemical resistance
  • Analogue temperature control