Disposable particle filtering respirators, VFlex™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
111-0671EA 0 DKK
111-0671
Disposable particle filtering respirators, VFlex™
Åndedrætsværn Engangsmasker Partikelmasker
This lightweight and flexible disposable FFP2 half mask provides effective respiratory protection against dust and mist (up to 12×TLV for particulates) at low costs. The V-shaped pleats flex and expand with the mouth movement and therefore enable easier talking and breathing. Positioning tabs, the adjustable nose clip and a choice of two different sizes also help to achieve a custom fit.
  • Embossed front panel helps the respirator retain its shape away from the mouth
  • Spacious and comfortable inside, with minimal impact on field of vision
  • Flat-fold convenience – convenient storage prior to use
Certificeringer: EN149:2001; A1:2009 FFP2 NR D