Prøvebeholdere, til engangsbrug, med låg
Leverandør: Simport
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SIMPC570-12EA 3950 DKK
SIMPC570-12
Prøvebeholdere, til engangsbrug, med låg
Prøvecontainere Allround kasser
Graduated container (PP) with screw cap (PE). Heavy-duty thick wall construction of both container and lid ensures a positive leakproof seal.
  • Wide base design for stable reliable use
  • Molded-in graduations up to 128 ml
  • Non-sterile
The drip ring on the container reduces the chances of contamination.
Emballering: Packaged in bags of 100 stacked by 10's. Yellow caps packed separately in bags of 100.