Disinfectant wipes, Aseptil
Leverandør: Laboratoires Prodene Klint
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LBPKU1A84JI022EA 505 DKK
LBPKU1A84JI022
Disinfectant wipes, Aseptil
Papirservietter Engangsservietter og - håndklæder Desinficerende servietter
These wipes are impregnated with an antiseptic and disinfectant solution for the antisepsis of the hands and for the disinfection of objects, surfaces and medical appliances in a medical environment.
  • Made of a mixture of cellulose and viscose
  • The solution has been formulated to minimize the risk of allergic reactions
  • They might also be used during travelling, in airplanes, at the office or at home
Ingredients: Aqua, alcohol denat. CAS 64-17-5 (47 mg/g), propylene glycol, myristalkonium chloride CAS 139-08-2 (3 mg/g), menthol, chlorhexidine digluconate CAS 18472-51-0 (1,2 mg/g).
Wipes are effective against bactericides, fungicides and virucides.
Colour: White.
Leveringsinformation: Supplied in dispenser box: Wipes can easily be dispensed one by one.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904