These wipes are impregnated with an antiseptic and disinfectant solution for the antisepsis of the hands and for the disinfection of objects, surfaces and medical appliances in a medical environment.
- Made of a mixture of cellulose and viscose
- The solution has been formulated to minimize the risk of allergic reactions
- They might also be used during travelling, in airplanes, at the office or at home
Ingredients: Aqua, alcohol denat. CAS 64-17-5 (47 mg/g), propylene glycol, myristalkonium chloride CAS 139-08-2 (3 mg/g), menthol, chlorhexidine digluconate CAS 18472-51-0 (1,2 mg/g).
Wipes are effective against bactericides, fungicides and virucides.
Colour: White.
Leveringsinformation: Supplied in dispenser box: Wipes can easily be dispensed one by one.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904