Kolber, pæreformede med standardslib, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SCOT241950809EA 1850 DKK
SCOT241950809 SCOT241952004 SCOT241952501 SCOT241951402
Kolber, pæreformede med standardslib, DURAN®
Kolber Pæreformede flasker
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The conical geometry makes them ideal for small-scale reactions.