Heptadecafluor-1-octansulfonsyre

Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EHERC15987120EA 1060 DKK
EHERC15987120
Heptadecafluor-1-octansulfonsyre
Heptadecafluor-1-octansulfonsyre
Formel: C₈HF₁₇O₃S
Molekylvægt: 500,13 g/mol
MDL Number: MFCD00042454
CAS-nummer: 1763-23-1
EINECS: 217-179-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR