Wash bottles, wide neck, Chemware®
Leverandør: Saint-Gobain
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VERND1069742EA 10850 DKK
VERND1069742 VERND1069741 VERND1069740
Wash bottles, wide neck, Chemware®
PFA, translucent.
  • Chemically inert and leak-proof
  • Flexible nozzle with 360º swivel; small orifice allows accurate content dispensing
  • Transparent and break resistant
  • Molded-in graduation marks
These wash bottles feature a wide-mouth opening for convenience in filling and pouring of all types of liquids. Non-stick and easy to clean, fluoropolymer construction provides superior flex life and mechanical strength. As PFA contains the lowest concentration of trace metal ions of all fluoropolymers, these bottles are ideal for ultra-pure applications and environmental work.