Pose til prøvetagning af gas/væsker, Chemware®, Tedlar®
Leverandør: Saint-Gobain
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Chemware®
300-0280EA 812 DKK
300-0280 300-0288
Pose til prøvetagning af gas/væsker, Chemware®, Tedlar®
Poser Gasposer
Made from 2 mil thick PVF fluoropolymer film. When the chemical inertness of FEP is not required, Chemware® heat-sealed, PVF gas sampling bags are the ideal choice. These reusable bags are the perfect economical alternative for analysis of non-corrosive gases. Can be used when the user is dealing with carbon monoxide, sulphur dioxide, hydrogen sulfide, radon and mercaptans. Ideal for extracting air samples from smoke stack exhaust systems in fuel gas sampling applications.
  • Tough, easy to use, reliable
  • Highly flexible – easy to handle and store
  • Leak proof, heat sealed all four sides
  • No aging – unlimited storage life
  • Unique seam construction for added strength
  • Non-adhesive, nothing sticks
When the chemical inertness of FEP is not required, Chemware® heat-sealed, PVF gas sampling bags are the ideal choice. These reusable bags are the perfect economical alternative for analysis of non-corrosive gases. Just as in Chemware’s FEP bags, Chemware’s PVF gas sampling bags accept two fitting options – either a nickel-plated brass on/off valve, or a PTFE/silicone septum fitting in a polypropylene housing.Durable, heat-sealed seams ensure leak-proof operation.
Just as in Chemware®’s FEP bags, Chemware®’s PVF gas sampling bags accept two fitting options – either a nickel-plated brass on/off valve, or a PTFE/silicone septum fitting in a polypropylene housing. Durable, heat-sealed seams ensure leak-proof operation.