Kolonnehoved med ventilstophane
Leverandør: Lenz
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
271-3215EA 9090 DKK
271-3215
Kolonnehoved med ventilstophane
Destillations komponenter Hoveder
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With PTFE valve for easy and accurate reflux adjustment, with thermometer connection NS 14/23 for thermometers with 55 mm built-in length.