Flaskedispensere, med flaske
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRND43016EA 779 DKK
BRND43016
Flaskedispensere, med flaske
Dispenser head made of borosilicate glass 3.3, with inscriptions in green enamel.
  • Calibrated to deliver (TD, Ex)
  • Automatic dispenser particularly suited for dispensing suspensions
  • Complete with reservoir bottle 1000 ml capacity with neck size NS 29/32
Fejlgrænser: ± 1,0 ml (5/10 ml dispensere); ± 2,0 ml (20 ml dispensere); ± 2,5 ml (25 ml dispensere); ± 5,0 ml (50 ml dispensere); ± 10,0 ml (100 ml dispensere)