Dibutyldiphenyltin
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC12251200EA
912
DKK
EHERC12251200
Dibutyldiphenyltin
Dibutyldiphenyltin
Formel:
C₂₀H₂₈Sn Molekylvægt: 387,15 g/mol |
CAS-nummer:
6452-61-5 EINECS: 229-256-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...