2-Amino-5-nitro-6-picolin 98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

ACRO360770050EA 1820 DKK
ACRO360770050
2-Amino-5-nitro-6-picolin 98%
2-Amino-5-nitro-6-picolin
Formel: C₆H₇N₃O₂
Molekylvægt: 153,14 g/mol
Smeltepunkt: 185…189 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00053582
CAS-nummer: 22280-62-2
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Yellow to beige to brown Powder
Infrared spectrum Conforms
Melting point 186 to 192 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR