Pasteur pipettes
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-1719EA 715 DKK
612-1719 612-1720
Pasteur pipettes
Pipetter Pasteur-/Overførselspipetter
Soda-lime-glass.
Long drawn-out capillary tip. Suction end with constriction for a cotton plug. int.Ø of the tip: 1,0 mm.
Certificeringer: ISO 7712