Undecanal
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC17896200EA
0
DKK
EHERC17896200
Undecanal
Undecanal
Formel:
C₁₁H₂₂O Molekylvægt: 170,3 g/mol Kogepunkt: 226 °C (1013 hPa) Smeltepunkt: –2 °C Massefylde: 0,827 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007016 CAS-nummer: 112-44-7 EINECS: 203-972-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...