Hydrogenperoxid 30% (w/v) (100 volumes), ARISTAR® til sporstofanalyse

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

452024TEA 0 DKK
452024T
Hydrogenperoxid 30% (w/v) (100 volumes), ARISTAR® til sporstofanalyse
Hydrogen peroxide
Formel: H₂O₂
Molekylvægt: 34,01 g/mol
Kogepunkt: ∼107 °C (1013 hPa)
Smeltepunkt: < 0 °C
Massefylde: 1,11 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011333
CAS-nummer: 7722-84-1
EINECS: 231-765-0
UN nummer: 2014
ADR: 5.1,II

Bestil nu

Specifikationer Testresultater

Assay Min 30.0 % p/v
Acidity (as H2SO4) Max 30 ppm
Phosphate (PO4) Max 2 ppm
Sulphates (SO4) Max 2 ppm
Chloride (Cl) Max 0.5 ppm
Ag (Silver) Max 0.001 ppm
Al (Aluminium) Max 0.005 ppm
Au (Gold) Max 0.005 ppm
Ba (Barium) Max 0.005 ppm
Be (Beryllium) Max 0.001 ppm
Bi (Bismuth) Max 0.002 ppm
Ca (Calcium) Max 0.02 ppm
Cd (Cadmium) Max 0.001 ppm
Co (Cobalt) Max 0.002 ppm
Cr (Chromium) Max 0.001 ppm
Cu (Copper) Max 0.001 ppm
Fe (Iron) Max 0.01 ppm
Ga (Gallium) Max 0.005 ppm
In (Indium) Max 0.002 ppm
K (Potassium) Max 0.005 ppm
Li (Lithium) Max 0.001 ppm
Mg (Magnesium) Max 0.005 ppm
Mn (Manganese) Max 0.001 ppm
Mo (Molybdenum) Max 0.001 ppm
N (Nitrogen) Max 4 ppm
Na (Sodium) Max 0.02 ppm
Ni (Nickel) Max 0.002 ppm
Pb (Lead) Max 0.005 ppm
Sn (Tin) Max 0.001 ppm
Sr (Strontium) Max 0.001 ppm
Ti (Titanium) Max 0.005 ppm
Tl (Thallium) Max 0.001 ppm
V (Vanadium) Max 0.001 ppm
Zn (Zinc) Max 0.005 ppm
Zr (Zirconium) Max 0.001 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR