Vaskeflasker, smal åbning, med skruelåg
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRND143848EA 1370 DKK
BRND143848 BRND143862 BRND143854
Vaskeflasker, smal åbning, med skruelåg
Flasker Sprøjteflasker
PFA, translucent, with ETFE screw cap and FEP spray tube.
  • Excellent chemical resistance
  • High thermal stability from −200 to 260 °C
  • Can be used for less critical applications for which pure PFA is not necessary
With recycled PFA content. Thus, reasonably priced and environmentally friendly.
  • Excellent chemical resistance
  • High thermal stability from -200 °C to +260 °C
  • Can be used for less critical applications for which pure PFA is not necessary
With recycled PFA content. Thus, reasonably priced and environmentally friendly.