Funnels for vacuum filtration
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
516-7580EA 2810 DKK
516-7580 516-7579
Funnels for vacuum filtration
Tragte Filtreringstragte Filtertragte
Polysulfone, transparent.
  • Transparent funnels with graduations permit easy visual measurement
  • Less expensive than stainless steel and more durable than glass
  • Changing filters is easy
  • Twist-lock coupling minimises filter tearing
Ideal for liquid clarification, vacuum filtration, and scintillation counting studies.