GFL distilled water apparatus is used in research and development when preparing bacteriological and clinical samples, as well as for the preparation of cell and tissue cultures and the formulation of reagents and ointments. The distillate is also used for cleaning and sterilising, for buffer solutions and for microbiological and analytical applications. For the production of single distilled water (approx. 2,3 µs/cm).
- With thermometer to check the cooling water
- Thermostatic safety (dry run protection by thermostat)
- Energy saving by distillation of preheated cooling water
- Evaporator easily accessible for cleaning
- CO₂ degassing by the condenser
Water heater body, boiler, tank and stainless steel condenser, zinc-plated stainless steel double envelope housing with electrostatic epoxy powder coating.
Certificeringer: The distillate meets the standards of the international pharmacopoeia.
Bestillingsoplysninger: Supply hoses and water outlet hoses supplied as an option.