Vanddestillationsapparat, Merit® W4000
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
stuart®
171-1207EA 13460 DKK
171-1207 171-1201
Vanddestillationsapparat, Merit® W4000
Vandrensningssystemer Destillations apparatur
The Merit® W4000 water still produces up to 4 Litres per hour of distilled water. It incorporates a glass boiler and high efficiency condenser which provides exceptionally high quality pyrogen-free distillate (although distillate should be tested). Screw threads are incorporated on all water connections so hoses can be fitted and removed easily and safely without risk of breakage. The metal heater is fitted with a thermal cut-out to protect the still in the event of water failure. A built-in funnel and stopcock allow the still to be cleaned easily without hazardous dismantling.
  • Destillatet afkøles af den højeffektive spiralkondenser
  • To uafhængige sikkerhedstermostater
  • Nem rengøring uden adskillelse
  • Kan monteres på væg eller bord
Vanddestillationsapparatets varmeenheder er undtaget fra de 3 års garanti og er dækket indtil 1000 timers brug.