Microplate shakers, lab scale, SSL5
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
444-1628EA 22650 DKK
444-1628
Microplate shakers, lab scale, SSL5
Rystere Mikropladerystere
Designed for use with microtitre plates and microcentrifuge tubes. The aluminium tray is fitted with a specially designed mat that securely holds up to eight microwell plates in place. High speed combined with a tiny orbit creates the vibrational shaking action required for mixing the very small volumes contained in the wells or in microcentrifuge tubes. Speed and time is easily set via the digital display.
  • Digital selection of mixing speed and time
  • Digital timer with alert allows a process time in hours, minutes or seconds or continuous mode
  • Ideal for use under controlled conditions, can be used at 4 to 40 °C, in an incubator or CO₂ incubator
  • BioCote®  antimicrobial protection
Certificeringer: IP Protection Class according to DIN EN 60529: IP 31
Bestillingsoplysninger: Tube holders are available as optional accessories. They fit securely onto the platform turning the unit into an effective tube mixer.