Luxmeter, testo 545
Leverandør: Testo
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0186EA 3680 DKK
TEST0560.0545
Luxmeter, testo 545
In order to have a good quality light, the luminous intensity in workplaces, hospitals, offices and schools has to fulfill specific minimum values. This can be measured and documented efficiently using the testo 545. A location list with individual luminous intensity values can be saved and later connected to form a curve, using a software. Light meter with location management.
  • Multi-point or timed mean calculation
  • Stores up to 99 file locations
  • Logger function (3000 readings)
  • Quick documentation on site
  • LCD display on four lines each 13 mm
Bestillingsoplysninger: Log printer available separately.
Leveringsinformation: Supplied with probe, 9 V battery for an operating life of 50 hours, and calibration protocol.