Flame photometers, PFP7 and PFP7/C
Leverandør: Cole Parmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
JENW83055-02EA 62400 DKK
JENW83055-02 633-0208 633-0209
Flame photometers, PFP7 and PFP7/C
Spektrofotometer og fotometer Flammefotometre
PFP7 are low temperature flame photometers designed for the determination of sodium and potassium in industrial applications – calcium, barium and lithium are optional.
  • Electronic ignition with detection system for user safety
  • Fine and coarse sensitivity controls on propane, butane, natural gas or LPG
  • Readings of both sodium and potassium at normal serum concentrations (mmol/L)
Bestillingsoplysninger: An oil- and moisture-free compressed air supply, 6 L/min at 1 kg/cm² together with a gas supply must be ordered separately.
Leveringsinformation: Supplied with filters (Na, K, Li, Ba, Ca (PFP7/I) and Na, K, Li (PFP7/C)). Both models are supplied with connecting hoses and clips, compressor plug, drain tap and operating instructions.
Advarsel: Serum samples must be diluted 200:1 or 100:1 prior to presentation.