Du Søgte efter: Rotationsfordampere
20  results were found
SearchResultCount:"20"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: VWR Collection
Beskrivelse: This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional...
Tilbud
Leverandør: VWR Collection
Beskrivelse: This is an essential instrument for use in chemical laboratories for efficient and gentle removal of solvents from samples by evaporation. It offers a...
Leverandør: Heidolph Instruments GmbH & Co.KG
Beskrivelse: Hei-VAP Expert Control rotary evaporators allow central control of all process parameters: Vacuum, cooling temperature, rotation and heating bath temp...
Leverandør: Heidolph Instruments GmbH & Co.KG
Beskrivelse: These evaporators are equipped with proven smart details such as Easy-Clip, patented clamping sleeve, lift stop to limit immersion depth and universal...
Leverandør: Heidolph Instruments GmbH & Co.KG
Beskrivelse: Industrifordampere, til 20 l.
Leverandør: IKA
Beskrivelse: Rotationsfordampere med indbygget varmebad, sikker og enkel betjening ved hjælp af ergonomiske knapper på forsiden.
Leverandør: IKA
Beskrivelse: These entry-level evaporators are ideal for all standard evaporation applications in universities and research laboratories. The functions of the RV10...
Varenummer: (IKAA10012324)
Leverandør: IKA
Beskrivelse: The RV 3 pro V-C Complete package is the complete solution for your rotary evaporator setup as all components and connections required for operation a...
UOM: 1 * 1 Items
New Product
Varenummer: (181-0471)
Leverandør: KNF
Beskrivelse: These two evaporators were designed to provide reliable daily performance in different laboratory environments.
UOM: 1 * 1 Items
Varenummer: (531-1453)
Leverandør: IKA
Beskrivelse: The RV 10 auto pro V-C Complete package is the complete solution for your rotary evaporator set-up. In addition to the rotary evaporator RV 10 auto, t...
UOM: 1 * 1 Items
Leverandør: IKA
Beskrivelse: Rotary evaporators with integrated heating bath, features include an innovative control concept with a bright, high-contrast, TFT, colour-graphic cont...
Leverandør: IKA
Beskrivelse: The RV 3 rotary evaporator is the ideal entry-level model from the IKA rotary evaporator portfolio. It is suitable for a multitude of uses in the chem...
Leverandør: BUCHI
Beskrivelse: This ATEX-compliant version of the R-220 was specially developed for applications in EX zones and is compliant with the extremely stringent ATEX 95 di...
Leverandør: BUCHI
Beskrivelse: Its modular design allows the extension of the Rotavapor® R-300 to a fully integrated system where a central interface regulates each component.
Leverandør: BUCHI
Beskrivelse: The R-100 is available as a stand-alone unit or as a complete ready to use kit (Essential Solution kit). The Essential Solution kits provide an econom...
Leverandør: BUCHI
Beskrivelse: The Rotavapor R-250 EX is the EX rated version of the Rotavapor R-250. It fulfills the high requirements of the ATEX regulations and fits perfect to y...
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 20
no targeter for Bottom