Combined Kjeldahl mineralisation and distillation ramps
Leverandør: Gerhardt
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
705-0480EA 65350 DKK
705-0480 705-0479
Combined Kjeldahl mineralisation and distillation ramps
Destillationssystemer Kjeldahl-destillationssystemer
Two level apparatus for six mineralisations and six distillations.
  • Heating produced by a six-hotplate ramp
  • Mineralisation ramp with run-off chamber
  • Up to 650 °C for the type KI13/26 2×400 V, 3 phase, power 6960 W
W×D×H: 970×470×1820 mm
Leveringsinformation: Delivered with run-off tubes, fittings, water cell and water aspirator.