Temperaturtabelskriver, KT800
Leverandør: Dickson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2246EA 9610 DKK
DICKKT8P0 DICKKT8P2 DICKKT8P3
Temperaturtabelskriver, KT800
This chart recorder on Ø 203 mm disc provides more flexibility, a flip-up pen arm for fast pen and chart replacement and a digital display (620-2247 only). Remote K-thermocouple probe with replaceable sensor.
  • User selectable temperature ranges and recording times
  • Recording time settable: 24 hours, 7 and 31 days
  • Audible and visual alarms
Leveringsinformation: Supplied with four AA batteries and mains supply, a red pen and a quick start guide. Charts (203 mm) need to be ordered separately.