These tissues have been bleached without chlorine and are biodegradable. They can be used to quickly disinfect alcohol-resistant surfaces.
Composition:
One Pursept® A tissue contains 1027 mg ethanol, 0,56 mg n-alkyl-aminopropyl-glycine, auxiliary agents.
Physical and chemical properties:
Active solution (Pursept® A Xpress): clear, colourless liquid, with pleasant, fresh smell
Density: ±0,9 g/ml
pH: ±7,5
Microbiological action/ Concentration:
Bacteria and fungi with little organic burden with mechanics to DGHM/VAH: conc. / 15 seconds
Limited virucidal efficacy (incl. HIV, HBV, HCV): conc. / 15 seconds
Efficacy against TbB: conc. / 30 seconds
Advarsel: It is recommended to read the notes and information regarding this product before each use.