VWR® ND8, krympehalshætteglas
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
548-0036AEA 2140 DKK
548-0036A 548-0035A 548-0037A 548-0035 548-0036 548-0340A 548-0341A 548-0078A 548-0783A 548-0831A 548-0339A 548-0080A
VWR® ND8, krympehalshætteglas
Hætteglas
These vials are for use on instruments from the following manufacturers: Agilent, Beckman, Carlo Erba, CTC, Fisons, PerkinElmer, Shimadzu, Thermo Scientific, VWR®/Hitachi, more.
  • Flat, round or conical bottom
  • In clear or amber glass
  • For almost all autosamplers
Broad selection of crimp neck vials ND8 available: crimp neck vials and micro vials ND8 can be closed with 8 mm aluminium caps, 9 mm PE caps or with 8 mm push-on caps. However, micro vials often need an adapter to run in the autosampler. They often have a conical bottom shape, so that they cannot stand by themselves, but need an adapter.
  • With flat, round or conical bottom
  • In clear or amber glass
  • For almost all autosamplers
Broad selection of crimp neck vials ND8 available: Crimp neck vials and micro-vials ND8 can be closed with 8 mm aluminium caps, 9 mm PE caps or with 8 mm push-on caps. However, micro-vials often need an adapter to run in the autosampler. They often have a conical bottom shape, so need an adapter to stand.