VWR® SuperClear, Ultra-High Performance Centrifuge Tubes
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
525-0156EA 1620 DKK
525-0156 VWRU21008-103 525-0155 525-0151 VWRU21008-089 VWRU21008-178 525-0150 525-0306 VWRU21008-195 21008-242 VWRU82018-048 21008-240 21008-214 21008-216 21008-105 VWRU93000-032 82018-050
VWR® SuperClear, Ultra-High Performance Centrifuge Tubes
Rør Centrifugerør
These popular, conical-bottom centrifuge tubes are designed for high speed centrifugation, and are made of non cytotoxic, medical grade resin that allows users to easily see sample volume and colour.
  • Prevents breakage and leakage during high speed centrifugation
  • Easy to read graduations for accurate measurements
  • Patented extra large, smear resistant writing panel
  • Versatile packaging options
  • Free from RNase, DNase, endotoxin and ATP
SuperClear tubes feature highly visible printed graduations for easy filling and sample preparation. The patented, extra large writing panel is the largest on the market, and transparent for easy viewing of samples through the label. This writing panel is also smear resistant for permanent coding that won’t wear off when inserting and removing tubes from a centrifuge rotor.
Caps are available in two styles: flat and plug. Flat caps are leak resistant and feature a moulded-in elastomeric sealing ring that keeps the sealed tubes completely secure. These caps are ideal for vortexing chemicals and long rocker incubations. Plug caps feature a deep sealing area and offer a secure seal for extended storage.
Caps must be loosely attached to tubes when autoclaving.