Microarray slides, LifterSlips™
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
737-0015EA 0 DKK
MENZK822X25I24816
Microarray slides, LifterSlips™
LifterSlips™ are manufactured from glass chosen for flatness and superior surface quality.
Increased rigidity reduces variation in hybridisation fluid volume.