Safety shield, single piece
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-1308EA 5180 DKK
AZLOSGT102
Safety shield, single piece
Very strong and lightweight safety shield made from PC (transparent, thickness: 4 mm).
  • Excellent protection in potentially explosive situations
  • Stand can be clamped or bolted onto the bench to give greater stability
Leveringsinformation: Supplied with a pair of plastic coated metal feet.