Pipettehjælper, macro
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-1935EA 671 DKK
612-1935 612-1932 612-1900 612-1933
Pipettehjælper, macro
Pipetter Pipettehjælpere
Macro pipette controller for glass and plastic pipettes from 0,1 to 200 ml.
  • Conical silicone adapter offers a secure fit for different diameters
  • Valve system allows easy compression of the bellows
  • Spring-loaded lever enables sensitive filling and delivery of liquids
  • Meniscus is easily adjustable
  • Fully autoclavable at 121 °C (2 bar)
Each pipette controller is supplied with a 3 μm spare membrane filter.