Adhæsionsobjektglas, SuperFrost® Ultra Plus, Menzel Gläser
Leverandør: Epredia
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-0099EA 270 DKK
MENZJ3800AMNZ
Adhæsionsobjektglas, SuperFrost® Ultra Plus, Menzel Gläser
Positive electrostatic attraction strongly binds frozen tissue sections for in-situ hibridisation.
  • Ground edges frosted end for labelling
  • Formaldehyde-fixed paraffin or resin tissue sections
  • Improved reproducibility of diagnostic staining processes