Du Søgte efter: Hjelme
92  results were found
SearchResultCount:"92"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: UVEX
Beskrivelse: Ultra-modern design, innovative variable ventilation system, PE safety shell with drainage channel, fabric cap with six fastening points with sliding ...
Leverandør: UVEX
Beskrivelse: These ventilated helmets provide protection even in extremely low temperatures (down to −30 °C). They also provide protection against splashes of molt...
Leverandør: UVEX
Beskrivelse: Disse ventilerede hjelme yder beskyttelse selv ved ekstremt lave temperaturer (ned til −30 °C). De yder også beskyttelse mod stænk af smeltet metal.
Leverandør: JSP LTD
Beskrivelse: One of the lightest full shell bump caps available on the market.
Leverandør: SCOTT SAFETY
Beskrivelse: The Style 300 is a stylish, lightweight and comfortable safety helmet design. Manufactured from UV stabilised, high density polyethylene and available...
Leverandør: UVEX
Beskrivelse: Airwing B-WR safety helmet made from a polyethylene shell.
Varenummer: (111-1913)
Leverandør: UVEX
Beskrivelse: This lightweight bump cap offers maximum wearer comfort and protection against bumps and grazes. The comfortable interior consists of four layers: Sho...
UOM: 1 * 1 Items
Varenummer: (MMMAG2C)
Leverandør: 3M
Beskrivelse: 3M™ pinlock suspension, G2C, is the replacement pinlock suspension cradle and plastic sweatband for all 3M™ hard hats (except the X5000 series).
UOM: 1 * 1 Items
Leverandør: JSP LTD
Beskrivelse: Visor carrier clips into universal slot ear defender housing for the Evolution® safety helmet range. Available with polycarbonate or acetate visors to...
Varenummer: (MMMAG2N)
Leverandør: 3M
Beskrivelse: 3M™ Ratchet suspension, G2N is a replacement ratchet suspension cradle and plastic sweatband for all 3M™ Hard Hats, except the X5000 series.
UOM: 1 * 1 Items
Leverandør: JSP LTD
Beskrivelse: EVO® VISTAlens® is a next generation, feature-rich safety helmet based on the proven Evolution® head protection technology. Incorporating a fully retr...
Leverandør: UVEX
Beskrivelse: uvex pheos letvægtssikkerhedshjelmen har et sporty design og giver bæreren maksimal komfort. Den robuste og stive hjelmskal modstår også nemt presset ...
Leverandør: UVEX
Beskrivelse: Lightweight helmets made from HDPE.
Leverandør: UVEX
Beskrivelse: This light ABS helmet offers a high level of protection and durability. It is multifunctional and combines features from industrial protection helmets...
Varenummer: (UVEX9772.544)
Leverandør: UVEX
Beskrivelse: This safety helmet is equipped with slots for uvex visors and earmuffs.
UOM: 1 * 1 Items
Leverandør: UVEX
Beskrivelse: These caps in fluorescent yellow or fluorescent orange feature an innovative hard shell element for reliable impact protection.
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 92
no targeter for Bottom