Glass tube cutter
Leverandør: Usbeck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201-9030EA 165 DKK
201-9030 201-9031
Glass tube cutter
Knive
Zinc diecast, nickel-plated with steel wheel.
  • For glass tubes up to 30 mm in diameter
  • Replaceable steel wheel
  • Easy handling