Du Søgte efter: Filtrerpapir
159  results were found
SearchResultCount:"159"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: VWR Collection
Beskrivelse: Grade 402 and 302: General grade for qualitative tasks. Suitable for most sulphides. Determination of total fat content in dairy products....
Varenummer: (516-0316)
Leverandør: VWR Collection
Beskrivelse: Low nitrogen content filters for determination of nitrogen content in steel and iron samples dissolved in acid. Filtration of fine precipitates to dis...
UOM: 1 * 100 Items
Leverandør: VWR Collection
Beskrivelse: 424: Bruges til filtrering af blysulfid, jernsulfid, sølvsulfid, alkalikarbonat, blychromat, arsenik, antimon, cadmium og fødevarer. Bestemmels...
Leverandør: VWR Collection
Beskrivelse: Hydrofobt filterpapir, der bruges til separation af vandige opløsninger og organiske opløsninger.
Leverandør: Whatman products (Cytiva)
Beskrivelse: Whatman Grade 589/2 filters from Cytiva are used in routine quantitative analyses, including analysis of aqueous suspension in paper, flour analysis,...
Leverandør: Whatman products (Cytiva)
Beskrivelse: Whatman Grade 589/3 ashless filter paper from Cytiva is particularly useful in capturing very fine particulate samples such as insoluble molecules in...
Leverandør: Whatman products (Cytiva)
Beskrivelse: These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also ava...
Leverandør: Whatman products (Cytiva)
Beskrivelse: Whatman Grade 1 Qualitative Filter Papers are the most widely used filter paper for routine laboratory applications, qualitative analytical separation...
Leverandør: Whatman products (Cytiva)
Beskrivelse: A prepleated filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration ...
Leverandør: Whatman products (Cytiva)
Beskrivelse: With less than 0,01% ash, Cytiva's Whatman Grade 589/1 filter papers can be used for the Blaine test in the cement industry and as an ash content tes...
Leverandør: Whatman products (Cytiva)
Beskrivelse: Den udstrakte anvendelse af Whatman kromatografipapir over hele verden afspejler produkternes renhed, høje kvalitet og konsistens. Papirerne er fremst...
Varenummer: (512-0636)
Leverandør: Whatman products (Cytiva)
Beskrivelse: Grade 181 support filter for seed tests using copenhagen tank system.
UOM: 1 * 100 Items
Leverandør: Whatman products (Cytiva)
Beskrivelse: Comparable to Grade 0858 but with a smooth surface, slightly thinner and faster.
Leverandør: Whatman products (Cytiva)
Beskrivelse: Cone folded filter paper saves valuable time when undertaking repetitive or multiple analyses.
Leverandør: Whatman products (Cytiva)
Beskrivelse: Whatman Grade 1V lab filter paper from Cytiva business is a widely used pre-folded filter for many general filtration applications.
Leverandør: Whatman products (Cytiva)
Beskrivelse: A general qualitative filter with creped surface. Widely used in industrial or student laboratories for routine filtration.
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 159
no targeter for Bottom