Du Søgte efter: Centrifugefiltre
45  results were found
SearchResultCount:"45"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: VWR Collection
Beskrivelse: Cyto strips made of filter board for cytocentrifuges. Absorption of excess liquids from the staining of samples in cytologic analysis. Can be used in ...
Leverandør: VWR Collection
Beskrivelse: PP-indsats og tilløbsrør. MWCO-filtre er ideelle til koncentrering, oprensning og afsaltning af peptider, proteiner, oligonukleotider, DNA og RNA.
Tilbud
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: These centrifugal devices can quickly concentrate up to 20 ml of biological sample without valuable sample loss.
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: These devices are used for simple, reliable concentrating and desalting of 50 to 500 μl samples. Select the low protein binding Omega™ membrane for ul...
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: Filter, Spin, Centrifugal, Microsep™, Advance, Membran: Omega™, MWCO: 1 kD
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: The Pall Nanosep device for nucleic acid binding (NAB) incorporates a dual layer silica-based quartz glass fiber media to allow for efficient binding ...
Tilbud
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: Convenient and reliable concentration, purification and diafiltration of 15 to 60 ml biological samples. Concentrate 60 ml sample volumes to 4 ml in 3...
Leverandør: Biotium
Beskrivelse: Ultrafiltration can be used to rapidly and easily remove small molecules such as Tris, glycerol and glycine from antibody solutions, or to concentrate...
Leverandør: Cytiva (Formerly Pall Lab)
Beskrivelse: These centrifugal devices are suitable for rapid sample prepration in many applications.
Tilbud
Leverandør: G-Biosciences
Beskrivelse: These disposable spin columns are designed for protein purification. These spin columns are uniquely designed, with a snap off end converting to a clo...
Leverandør: G-Biosciences
Beskrivelse: These C18 spin columns are ready-to-use micro centrifuge columns for peptide clean up and concentration. The columns consist of porous C18 reverse-pha...
Varenummer: (AGLS5188-5202)
Leverandør: AGILENT
Beskrivelse: Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.
UOM: 1 * 25 Items
Leverandør: Sartorius
Beskrivelse: Vivaspin® concentrators are disposable ultrafiltration devices for the concentration of biological samples of 0,4 to 2 ml.
Leverandør: Sartorius
Beskrivelse: Vivaspin® centrifugal concentrators are disposable ultrafiltration devices for the concentration of biological samples. Vivaspin® 500 centrifugal filt...
Leverandør: Sartorius
Beskrivelse: Vivaspin® Turbo 4 can handle 0,04 to 4 ml sample volume in swing-out and fixed angle rotors accepting 15 ml centrifuge tubes.
Leverandør: Sartorius
Beskrivelse: Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in eithe...
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 45
no targeter for Bottom