Du Søgte efter: Høreværn
49  results were found
SearchResultCount:"49"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: JSP LTD
Beskrivelse: The Sonis® ear defender range has been designed with a colour coded system for simple selection, and peaks at an unbeatable 37 SNR.
Leverandør: 3M
Beskrivelse: Optime™ II earmuffs are available in various designs for different requirements: With foldable headband, neckband or as a helmet attachment.
Leverandør: 3M
Beskrivelse: Peltor™ LiteCom Headsets are hearing protectors with a built-in, pre-programmed two-way communication radio and an integrated noise cancelling microph...
Leverandør: 3M
Beskrivelse: The Peltor™ WS™ LiteCom Plus Headset is available in multiple mounting styles, and features an integrated pre-programmed analog two-way radio, Bluetoo...
Leverandør: 3M
Beskrivelse: Peltor™ X-serien af høreværn kombinerer fremragende beskyttelse med ultimativ komfort. De fås i bøjle- og hjelmmonterede versioner med en bred vifte a...
Varenummer: (MMMA7100123157)
Leverandør: 3M
Beskrivelse: Left and right side attachment conversion kit to earmuffs to Comtac XP and Comtac XPI headsets. Twin headband design provides heat dispersion and inte...
UOM: 1 * 10 Items
New Product
Varenummer: (MMMAX5P5E)
Leverandør: 3M
Beskrivelse: 3M™ PELTOR™ X5 Earmuffs are innovative hearing protectors that offer exceptional performance, without the need for double protection. Offering the sam...
UOM: 1 * 1 Items
Leverandør: 3M
Beskrivelse: Disse høreværn er udviklet til brug i miljøer med ekstremt høj støj, især miljøer, som er domineret af lavfrekvente lyde. Beskyttelsen er baseret på e...
Leverandør: 3M
Beskrivelse: Hygiejnesæt til høreværn Peltor™ Optime™ III
Leverandør: 3M
Beskrivelse: 3M™ PELTOR™ WS™ LiteCom Pro III headsets are high-performance hearing protectors with a built-in analogue and digital programmable two-way communicati...
Varenummer: (MMMA7100123139)
Leverandør: 3M
Beskrivelse: 3M™ PELTOR™ X1 earmuffs are our lightweight hearing protectors with comfortable, soft ear cushions. They feature an electrically insulated, twin headb...
UOM: 1 * 10 Items
New Product
Leverandør: 3M
Beskrivelse: ProTac™ III hearing protector helps protect against harmful noise, while allowing ambient sounds to be heard at a level below 82 dB. It increases the ...
Leverandør: 3M
Beskrivelse: Rechargeable battery pack for use with specific 3M™ Peltor™ headsets.
Varenummer: (111-2215)
Leverandør: 3M
Beskrivelse: These earmuffs offer a smooth, level-dependent hearing protection function that electronically cuts off abrupt, loud sound. They rapidly detect loud i...
UOM: 1 * 1 Items
Leverandør: 3M
Beskrivelse: The lightweight Peltor™ Optime™ I earmuffs provide a moderate level of attenuation that meets the needs of the majority of industrial applications.
Varenummer: (MMMAMRX21A3WS6)
Leverandør: 3M
Beskrivelse: The Peltor™ WS™ Alert™ XPI Headset is a is high-attenuating hearing protector that can connect up to two Bluetooth® enabled devices, such as a mobile ...
UOM: 1 * 1 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 49
no targeter for Bottom