Prøverør med skørt
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
16465-262
Prøverør med skørt
Prøvecontainere Allround kasser
PP tubes with HDPE screw cap.
  • Robust, durable and leakproof
  • Sustainable design based on virgin resins
  • Wide skirt ensures stability when standing
  • Large, matt area for easy labelling and identification
Leakproof freestanding mailing tubes with screw caps feature graduations at 1 ml increments and a large frosted writing surface for convenient labeling and identification.
Certificeringer: Lab tested and certified to the IATA 95kPA standard for safe shipping of samples.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904.