Du Søgte efter: Tabeller
33  results were found
SearchResultCount:"33"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: Industrial Physics
Beskrivelse: Test charts are used for testing physical properties of coating, lacquers, and inks.
Varenummer: (761-0477)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Periodic table, wall poster. New Swedish redesigned wall poster in strong specially treated fabric with sewn channels at the top and bottom. Exciting ...
UOM: 1 * 1 Items
Varenummer: (766-0260)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: Reproduces in multicolor printing a number of different absorption and emission spectra including solar spectrum with the main Fraunhofer lines and th...
UOM: 1 * 1 Items
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: With colorful pictures. 100×70 cm of durable plastic-coated paper.
Varenummer: (763-0208)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: The 'bud cloth' makes it possible to reach the right tree and shrub even in winter.
UOM: 1 * 1 Items
Varenummer: (763-0204)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: How many legs? Cover or not? Small animals on the beach can be identified within text and image.
UOM: 1 * 1 Items
Varenummer: (763-0209)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: About the most common fungi, the determination key gives information. Clear markings for edible (Ä), inedible (OÄ) and toxic fungi (G).
UOM: 1 * 1 Items
Varenummer: (763-0206)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: To which group do small animals in and by lakes and rivers belong? With the text and image of the determination key, you can find your way around.
UOM: 1 * 1 Items
Varenummer: (763-0207)
Leverandør: EDUCATIONSCIENCE COLLECTION P
Beskrivelse: The 'leaf cloth' determination key helps to identify about 25 common trees and shrubs.
UOM: 1 * 1 Items
Varenummer: (116-0424)
Leverandør: SAFETY FIRST AID GROUP LTD
Beskrivelse: The Chemical Spills poster has clear step by step instructions on how to react in case of a chemical spill and how to provide treatment to someone tha...
UOM: 1 * 1 Items
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: These colourful anatomical charts depict various parts of the human body.
Varenummer: (763-0162)
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: Plank mounted on roll. 84×118 cm.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (763-0165)
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: Anatomical poster suitable for wall mounting of the three types of common muscle tissue. 840×1180 mm.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (763-0150)
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: This poster is mounted on a reel.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (763-0157)
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: This poster is mounted on a reel.
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Varenummer: (761-0013)
Leverandør: Binhold, Paul Lehrmittel
Beskrivelse: This colourful large wall chart illustrates the different stages of cellular division, meiosis. The mammalian cells on the chart are coloured accordin...
UOM: 1 * 1 Items
Market Source Item This is a MarketSource item. Additional charges may apply
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 33
no targeter for Bottom