Sprøjteflasker, smal hals med påtrykt navn, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Nalgene®
215-0981EA 302 DKK
215-0981 215-0980 215-0983
Sprøjteflasker, smal hals med påtrykt navn, Nalgene®
Flasker Sprøjteflasker
LDPE, translucent, PP spray cap.
  • To allow easy hazard and chemical identification, as well as to prevent cross-contamination, each bottle has a colour bar that corresponds to its coloured closure
  • Non metallic vent prevents solvent leakage
  • Each bottle features chemical name, ICS chemical formula, CE index number and CAS number
  • Uses GHS compliant symbols
All bottles are clearly printed with hazard type and chemical name in high quality, chemical resistant inks.