Du Søgte efter: Opbevaringsposer
31  results were found
SearchResultCount:"31"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: VWR Collection
Beskrivelse: 3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropel...
Leverandør: VWR Collection
Beskrivelse: VWR® slider zip bags, in LDPE, transparent, with blue slider. For the packing and storage of goods.
Tilbud
Varenummer: (BRDY065780)
Leverandør: Brady
Beskrivelse: Lockout bag made of rugged canvas with drawstrings.
UOM: 1 * 1 Items
Leverandør: Flexico
Beskrivelse: The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good ba...
New Product
Leverandør: DESCO
Beskrivelse: Opbevaringsposer, Metal-In shielding bubble bags with 50 mm flap, Mål: 355×455 mm
Leverandør: DESCO
Beskrivelse: Transparent metal-in static shielding bag provides a static safe environment for ESD sensitive electronic devices (ESDS).
Varenummer: (DESCPCL1001115)
Leverandør: DESCO
Beskrivelse: Metal-In static shielding bags are manufactured from a polyester, metal, polyethylene laminate. ESD safe packaging designed for use in cleanroom areas...
UOM: 1 * 100 Items
Varenummer: (MINIITM080303)
Leverandør: Flexico
Beskrivelse: Zip seal bags designed for food contact.
UOM: 1 * 500 Items
New Product
Leverandør: DESCO
Beskrivelse: This aluminised moisture barrier bag is designed to provide packaging for ESD and moisture sensitive items, both inside and outside an ESD protected a...
Leverandør: Brady
Beskrivelse: These Lockout visit bag has several pockets to store and smoothly retrieve any lockout/tagout product.
Leverandør: NEW PIG
Beskrivelse: These bags are designe to protect, store and transport portable containment pools of different sizes.
Leverandør: BÜRKLE
Beskrivelse: Bags with the patented zipper are for ideal for storing, collecting, protecting and shipping of specimens, samples and sales products.
Leverandør: Nitritex
Beskrivelse: Manufactured in a cleanroom environment from ultra-clean low-density polythene. Because the material is free of additives, there is no contamination f...
Leverandør: DELTA T
Beskrivelse: BlueLine Bags are economical and durable. Its insulating property ensures the maintainance of constant internal temperature and protects the samples a...
Leverandør: DELTA T
Beskrivelse: SilverBags are the economical solution to the problem of short-term insulated transport of TempShells. The SilverBag range is made from tough nylon co...
Varenummer: (DAKL460698)
Leverandør: DAKLAPACK
Beskrivelse: Semi transparent PE bubble foil safety bag with closure type-destructive tape.
UOM: 1 * 100 Items
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 31
no targeter for Bottom