Du Søgte efter: Poser
VWR har et bredt udvalg af poser til laboratoriet eller generel anvendelse. Poser kan være special-lavet til, at kunne tåle farligt affald mærket med symbolet for farligt affald (Biohazard).
VWR har et bredt udvalg af poser til laboratoriet eller generel anvendelse. Poser kan være special-lavet til, at kunne tåle farligt affald mærket med symbolet for farligt affald (Biohazard).
303  results were found
SearchResultCount:"303"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Leverandør: Merck
Beskrivelse: The unique valve and seam-sealing process ensure that these bags are sturdy and virtually leak-proof, even under the most demanding sampling and shipp...
Leverandør: Merck
Beskrivelse: These screw cap valve (SCV) bags are suitable for calibration mixes - gas phase standards/gas mixtures.
Leverandør: VWR Collection
Beskrivelse: LDPE bags with safety tabs, manufactured and packed in sterile conditions.
Leverandør: VWR Collection
Beskrivelse: Zip bags in LDPE, for storage and transportation of solid samples, tubes, spare parts and documents, available with and without write-on panel.
Tilbud
Leverandør: VWR Collection
Beskrivelse: Wide choice of PE sterile bags from 80 to 3500 ml capacity. Fit all conventional laboratory blenders, adapted to all samples.
Leverandør: VWR Collection
Beskrivelse: This line of blue twist-seal printed bags, with safety tab and two round wires, has been specifically designed for food industry. The bags accommodate...
Leverandør: VWR Collection
Beskrivelse: Complete range of bags, sterile and alimentary certified, for safe and quick preparation of solid samples before microbiological and chemical analysis...
Leverandør: VWR Chemicals
Beskrivelse: This product is a quick and simple solution to collect microobiological samples in livestock housing, animal transportation vehicles, and their washin...
New Product Tilbud
Varenummer: (720-1985)
Leverandør: VWR Collection
Beskrivelse: Anti-tamper disposable bags, in PE, with double pocket for internal hospital biological sample transportation. Double pocket bags: Two separate compar...
UOM: 1 * 500 Items
Leverandør: VWR Collection
Beskrivelse: VWR® MEDBAGs are liquid-tight bags, used as internal hospital and external secondary packaging for the transportation of category B biological substan...
Leverandør: VWR Collection
Beskrivelse: PE bags, transparent. These sterile sampling bags provide a secure, contaminant-free pliant container that ensures dependable analysis results. Econom...
Leverandør: VWR Collection
Beskrivelse: LDPE bags with safety tabs, manufactured and packed in sterile conditions.
Leverandør: VWR Collection
Beskrivelse: These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
Tilbud
Leverandør: VWR Collection
Beskrivelse: 3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropel...
Leverandør: VWR Collection
Beskrivelse: Avantor single-use bioprocessing bags are designed to transfer and store your products throughout the drug manufacturing process. Made from proprietar...
Leverandør: VWR Collection
Beskrivelse: Robuste poser, som er ideel til deponering af biologisk affald, som skal autoklaveres først. Med wireaflukning, som tillader ekspansion af posen og ve...
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 303
no targeter for Bottom