Perchlorsyre 0.1 mol/l (0.1 N) i vandigopløsning Reag. Ph. Eur. 1062901
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
87886.180EA 393 DKK
87886.180
Perchlorsyre 0.1 mol/l (0.1 N) i vandigopløsning Reag. Ph. Eur. 1062901
Perchlorsyre
Bestillingsoplysninger: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formel: HClO₄
Molekylvægt: 100,46 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
UN nummer: 3264
ADR: 8,III
https://dk.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Bestil nu
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022