DNA fingerprinting, simu-gel, Ward's®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
732-1436EA 865 DKK
WARD148345
DNA fingerprinting, simu-gel, Ward's®
This simulated gel provides your students with actual results from WARD’S DNA Fingerprinting Lab Activity. The results are permanent, so it can be used at anytime to practice restriction mapping prior to performing the actual procedure or as a review for unit tests and final exams. The durable, flexible material simulates the look and feel of an agarose gel. It even contains wells so students can practice gel loading before using actual DNA.
  • W×L (mm): 60×100