Manometer
Leverandør: Gränna Plast-Emballage
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
765-0915EA 1050 DKK
GRAE192374-0
Manometer
Glass tube, wood base.
  • Graduations in centimetres
  • U-tube: Ext.Ø 10 mm, Int.Ø 7,5 mm, H 295 mm
  • Total height: 320 mm