Ethylcyanoacetat, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M-Clarity™ quality level = MQ200
8.00882.9025EA
0
DKK
8.00882.9999
8.00882.9025
Ethylcyanoacetat, Sigma-Aldrich®
Ethylcyanoacetat
Formel:
C₅H₇NO₂ Molekylvægt: 113,12 g/mol Kogepunkt: 209 °C (1013 hPa) Smeltepunkt: –22 °C Massefylde: 1,063 g/cm³ (20 °C) Flammepunkt: 104 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001940 CAS-nummer: 105-56-6 EINECS: 203-309-0 Merck Index: 13,03821 |
Specifikationer Testresultater
Solubility | 20 g/l (20°C) |
Melting Point | -22 °C |
Boiling Point | 209 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 110 °C |
Refractive Index | 1.4166 (20 °C, 589 nm) |
Density | 1.06 g/cm3 (20 °C) |
Ignition Point | 460°C |
Synonyms | Cyanoacetic acid ethyl ester |
Hill Formula | C5H7NO2 |
Chemical Formula | NCCH2COOC2H5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...