1,3,5-Trichlor-2-nitrobenzen
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC17761300EA
510
DKK
EHERC17761300
1,3,5-Trichlor-2-nitrobenzen
1,3,5-Trichlor-2-nitrobenzen
Formel:
C₆H₂Cl₃NO₂ Molekylvægt: 226,45 g/mol Smeltepunkt: 71 °C |
MDL Number:
MFCD00014690 CAS-nummer: 18708-70-8 EINECS: 242-518-1 UN nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...