Methyltrichloracetat
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
EHERC17684000EA
711
DKK
EHERC17684000
Methyltrichloracetat
Methyltrichloracetat
Formel:
C₃H₃Cl₃O₂ Molekylvægt: 177,41 g/mol Kogepunkt: 152 °C (1013 hPa) Smeltepunkt: –17,5 °C Massefylde: 1,4897 g/cm³ (20 °C) |
MDL Number:
MFCD00000794 CAS-nummer: 598-99-2 EINECS: 209-960-7 UN nummer: 2533 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...