2-Methylbenzofuran
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC15083785EA
1020
DKK
EHERC15083785
2-Methylbenzofuran
2-Methylbenzofuran
Formel:
C₉H₈O Molekylvægt: 132,16 g/mol |
MDL Number:
MFCD00005850 CAS-nummer: 4265-25-2 EINECS: 224-249-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...